Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

292761

Sigma-Aldrich

2-Hydroxyethyl acetate

50%, technical grade

Synonym(s):

2-Acetoxyethanol, Ethylene glycol monoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2OH
CAS Number:
Molecular Weight:
104.10
Beilstein/REAXYS Number:
1743114
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

technical grade

concentration

50%

impurities

<3% benzene

refractive index

n20/D 1.42 (lit.)

bp

187-189 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCCO

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains ethylene glycol and ethylene glycol diacetate

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service