Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

284831

Sigma-Aldrich

1-Nonadecene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CH=CH2
CAS Number:
Molecular Weight:
266.51
Beilstein/REAXYS Number:
1705613
EC Number:
MDL number:
UNSPSC Code:
12352100

assay

97%

refractive index

n20/D 1.445 (lit.)

mp

23 °C (lit.)

density

0.79 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC=C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service