Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

283533

Sigma-Aldrich

2,4-Dichloroiodobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3I
CAS Number:
Molecular Weight:
272.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.647 (lit.)

bp

262-263 °C (lit.)

density

2.01 g/mL at 25 °C (lit.)

functional group

chloro
iodo

SMILES string

Clc1ccc(I)c(Cl)c1

InChI

1S/C6H3Cl2I/c7-4-1-2-6(9)5(8)3-4/h1-3H

InChI key

ZQKJCBDCOGLKCQ-UHFFFAOYSA-N

General description

Ullmann coupling reaction of 2,4-dichloroiodobenzene with chlorinated iodobenzene has been investigated.

Application

2,4-Dichloroiodobenzene has been used in the preparation of 2,4,2′,4′ -tetrachlorobiphenyl, major component of commercial polychlorobiphenyls.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of sulfur-containing metabolites of 2,4,2',4'-tetrachlorobiphenyl in mice.
T Mizutani
Bulletin of environmental contamination and toxicology, 20(2), 219-226 (1978-08-01)
The Ullmann coupling reaction: A new approach to tetraarylstannanes.
Shaikh NS, et al.
Organometallics, 25(17), 4207-4214 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service