Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

281050

Sigma-Aldrich

Tricyclene

99%

Synonym(s):

1,7,7-Trimethyltricyclo[2,2,1,02.6]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein/REAXYS Number:
1901153
UNSPSC Code:
12352001
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99%

InChI

1S/C10H16/c1-9(2)6-4-7-8(5-6)10(7,9)3/h6-8H,4-5H2,1-3H3

InChI key

RRBYUSWBLVXTQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service