Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

278335

Sigma-Aldrich

Isobutyl isobutyrate

99%

Synonym(s):

2-Methylpropyl 2-methylpropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
144.21
Beilstein/REAXYS Number:
1701355
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

1 mmHg ( 39.9 °C)

assay

99%

autoignition temp.

810 °F

refractive index

n20/D 1.398 (lit.)

bp

145-152 °C (lit.)

mp

−81 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(=O)C(C)C

InChI

1S/C8H16O2/c1-6(2)5-10-8(9)7(3)4/h6-7H,5H2,1-4H3

InChI key

RXGUIWHIADMCFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service