Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

274178

Sigma-Aldrich

(1S,2S,5S)-(−)-Myrtanol

97%

Synonym(s):

(−)-trans-Myrtanol, (1S,2S)-10-Pinanol, (1S,2S)-6,6-Dimethylbicyclo[3.1.1]heptane-2-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein/REAXYS Number:
2322989
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

97%

optical activity

[α]20/D −28°, c = 4 in chloroform

refractive index

n20/D 1.489 (lit.)

bp

117-119 °C/15 mmHg (lit.)

density

0.966 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@H](CO)[C@]([H])(C1)C2(C)C

InChI

1S/C10H18O/c1-10(2)8-4-3-7(6-11)9(10)5-8/h7-9,11H,3-6H2,1-2H3/t7-,8+,9+/m1/s1

InChI key

LDWAIHWGMRVEFR-VGMNWLOBSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

186.8 °F - closed cup

flash_point_c

86 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service