Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

263559

Sigma-Aldrich

DL-Threitol

97%

Synonym(s):

DL-1,2,3,4-Butanetetrol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]2CH2OH
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

solubility

water: soluble 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

OC[C@@H](O)[C@H](O)CO

InChI

1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

UNXHWFMMPAWVPI-QWWZWVQMSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The manifestation of hydrogen bonding in the IR spectra of dl-threitol and erythritol (300-20 K).
Rozenberg M, et al.
Carbohydrate Research, 315(1), 89-97 (1999)
Hassina Massudi et al.
PloS one, 7(7), e42357-e42357 (2012-08-01)
Nicotinamide adenine dinucleotide (NAD(+)) is an essential electron transporter in mitochondrial respiration and oxidative phosphorylation. In genomic DNA, NAD(+) also represents the sole substrate for the nuclear repair enzyme, poly(ADP-ribose) polymerase (PARP) and the sirtuin family of NAD-dependent histone deacetylases.
Deoxygenation of biomass-derived feedstocks: oxorhenium-catalyzed deoxydehydration of sugars and sugar alcohols.
Mika Shiramizu et al.
Angewandte Chemie (International ed. in English), 51(32), 8082-8086 (2012-07-06)
Justyna Wojno et al.
ACS chemical biology, 7(5), 847-855 (2012-02-14)
Invariant natural killer T (iNKT) cells are restricted by the non-polymorphic MHC class I-like protein, CD1d, and activated following presentation of lipid antigens bound to CD1d molecules. The prototypical iNKT cell agonist is α-galactosyl ceramide (α-GalCer). CD1d-mediated activation of iNKT
J M Borggreven et al.
Caries research, 26(2), 84-88 (1992-01-01)
Some newly designed surface-active phosphates were investigated with respect to their capacity to inhibit caries lesion formation in vitro. On the labial or buccal surfaces of sound human third molars, windows were prepared for investigation. One half of each window

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service