Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

263400

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzyl chloride

98%

Synonym(s):

1-(Chloromethyl)-3,5-bis(trifluoromethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3CH2Cl
CAS Number:
Molecular Weight:
262.58
Beilstein/REAXYS Number:
656502
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

29-32 °C (lit.)

SMILES string

FC(F)(F)c1cc(CCl)cc(c1)C(F)(F)F

InChI

1S/C9H5ClF6/c10-4-5-1-6(8(11,12)13)3-7(2-5)9(14,15)16/h1-3H,4H2

InChI key

OINTXXMBRBLMHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Bis(trifluoromethyl)benzyl chloride has been used in the preparation of 1-(azidomethyl)-3,5-bis-(trifluoromethyl)benzene[1].

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved synthesis of 1-(azidomethyl)-3, 5-bis-(trifluoromethyl) benzene: development of batch and microflow azide processes.
Kopach ME, et al.
Organic Process Research & Development, 13(2), 152-160 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service