About This Item
Recommended Products
vapor pressure
96 mmHg ( 37.7 °C)
assay
96%
refractive index
n20/D 1.406 (lit.)
bp
95.7 °C (lit.)
density
0.703 g/mL at 25 °C (lit.)
SMILES string
[H]\C(CC)=C(/[H])CCC
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
19.4 °F - closed cup
flash_point_c
-7 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service