About This Item
Recommended Products
assay
97%
refractive index
n20/D 1.556 (lit.)
bp
231 °C (lit.)
density
0.925 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CC(=C)c1cccc(c1)C(C)=C
InChI
1S/C12H14/c1-9(2)11-6-5-7-12(8-11)10(3)4/h5-8H,1,3H2,2,4H3
InChI key
IBVPVTPPYGGAEL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1A
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
195.8 °F - closed cup
flash_point_c
91 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service