Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

254789

Sigma-Aldrich

1,10-Dichlorodecane

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)10Cl
CAS Number:
Molecular Weight:
211.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98%

refractive index

n20/D 1.4605 (lit.)

bp

167-168 °C/28 mmHg (lit.)

mp

15.6 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClCCCCCCCCCCCl

InChI

1S/C10H20Cl2/c11-9-7-5-3-1-2-4-6-8-10-12/h1-10H2

InChI key

RBBNTRDPSVZESY-UHFFFAOYSA-N

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Michel Wathier et al.
Journal of the American Chemical Society, 130(30), 9648-9649 (2008-07-03)
The synthesis of a supramolecular ionic network, its physical properties, and the use of this network property to form macroscopic porphyrin fibers are described. These ionic networks are compared to ionic liquids. Current ionic liquid compositions have a charge and
Photocatalytic degradation of 1, 10-dichlorodecane in aqueous suspensions of TiO2: a reaction of adsorbed chlorinated alkane with surface hydroxyl radicals.
El-Morsi TM, et al.
Environmental Science & Technology, 34(6), 1018-1022 (2000)
Nasser Goudarzi et al.
Journal of chromatographic science, 55(6), 669-675 (2017-03-24)
An improved microextraction method is proposed on the basis of ultrasound-assisted surfactant-enhanced emulsification and solidification of a floating organic droplet procedure combined with high performance liquid chromatography for the preconcentration and quantification of alprazolam (ALP) and chlordiazepoxide (CHL) present in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service