Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

235318

Sigma-Aldrich

Trimethyl 1,2,4-benzenetricarboxylate

98%

Synonym(s):

Trimethyl trimellitate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CO2CH3)3
CAS Number:
Molecular Weight:
252.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
solid
assay:
98%

vapor pressure

0.01 mmHg ( 20 °C)

assay

98%

form

solid

refractive index

n20/D 1.523 (lit.)

bp

194 °C/12 mmHg (lit.)

mp

38-40 °C (lit.)

density

1.261 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccc(C(=O)OC)c(c1)C(=O)OC

InChI

1S/C12H12O6/c1-16-10(13)7-4-5-8(11(14)17-2)9(6-7)12(15)18-3/h4-6H,1-3H3

InChI key

MJHNUUNSCNRGJE-UHFFFAOYSA-N

General description

Standard enthalpy of combustion of trimethyl 1,2,4-benzenetricarboxylate has been measured using bomb calorimetry.

Application

Trimethyl 1,2,4-benzenetricarboxylate has been used in the synthesis of polymers which can be cleaved at predetermined sites by ionizing radiations.

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

426.7 °F - Pensky-Martens closed cup

flash_point_c

219.3 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Standard enthalpies of formation of some methyl esters of benzene carboxylic acids.
Maksimuk YV, et al.
Journal of Chemical and Engineering Data, 43(3), 293-298 (1998)
Construction of polymer skeletons with radiation-scissile groups at predetermined sites.
Shimizu T, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 46(6), 1945-1953 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service