Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

235113

Sigma-Aldrich

Trimethylsilyl bromoacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CO2Si(CH3)3
CAS Number:
Molecular Weight:
211.13
Beilstein/REAXYS Number:
1927560
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

assay

98%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

57-58 °C/9 mmHg (lit.)

density

1.284 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OC(=O)CBr

InChI

1S/C5H11BrO2Si/c1-9(2,3)8-5(7)4-6/h4H2,1-3H3

InChI key

ZCHHFMWUDHXPFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylsilyl bromoacetate has been used:
  • in the preparation of new imidazolium salts functionalized with the trimethylsilyl ester group[1]
  • as alkylating agent in the synthesis of succinate fragment of pantocin B (antibiotic natural product)[2]
  • in the synthesis of taxol derivatives tethered at C2′ and C-7 to glutamate and folate[3]

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

116.6 °F - closed cup

flash_point_c

47 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper and palladium complexes with N-heterocyclic carbene ligands functionalised with carboxylate groups.
Danopoulos AA, et al.
Journal of Organometallic Chemistry, 693(21), 3369-3374 (2008)
Jae Wook Lee et al.
Bioorganic & medicinal chemistry, 10(7), 2397-2414 (2002-05-02)
A series of Taxol derivatives tethered at C2' and C-7 to glutamate and folate have been synthesized for evaluation as prodrugs which release Taxol via hydrolytic lability of their alpha-alkoxy and alpha-amino esters. The half-time for hydrolysis of these materials
The [1, 2]-thio-Wittig rearrangement: evidence for a radical mechanism and its suppression.
Bertolli P, et al.
Tetrahedron Letters, 47(45), 7939-7942 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service