Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

226068

Sigma-Aldrich

Allyloxytrimethylsilane

98%

Synonym(s):

3-(Trimethylsiloxy)propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2OSi(CH3)3
CAS Number:
Molecular Weight:
130.26
Beilstein/REAXYS Number:
1739517
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.397 (lit.)

bp

100-102 °C (lit.)

density

0.773 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OCC=C

InChI

1S/C6H14OSi/c1-5-6-7-8(2,3)4/h5H,1,6H2,2-4H3

InChI key

MNMVKGDEKPPREK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allyloxytrimethylsilane [3-(Trimethylsiloxy)propene] was used in the synthesis of trimethylsilyl protected (3-hydroxypropyl)-trimethoxysilane[1].

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

33.8 °F - closed cup

flash_point_c

1 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

John B Grande et al.
Dalton transactions (Cambridge, England : 2003), 39(39), 9369-9378 (2010-08-18)
The impressive surface activity of silicones can be enhanced by the incorporation of hydrophilic organic functional groups and polymers. Traditional routes to such compounds, which typically involve platinum-catalyzed hydrosilylation, suffer from incompatibility with certain functional groups. B(C(6)F(5))(3)-catalyzed condensation of hydrosilanes
Xiuhuan Song et al.
Polymers, 11(2) (2019-04-10)
A series of hybrid thermoplastic polyurethanes (PUs) were synthesized from bi-functional polyhedral oligomeric silsesquioxane (B-POSS) and polycaprolactone (PCL) using 1,6-hexamethylene diisocyanate (HDI) as a coupling agent for the first time. The newly synthesized hybrid materials were fully characterized in terms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service