Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

222674

Sigma-Aldrich

3-Fluoro-2-methylaniline

99%

Synonym(s):

3-Fluoro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)NH2
CAS Number:
Molecular Weight:
125.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

liquid

refractive index

n20/D 1.542 (lit.)

bp

89-91 °C/15 mmHg (lit.)

mp

7 °C (lit.)

density

1.099 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Cc1c(N)cccc1F

InChI

1S/C7H8FN/c1-5-6(8)3-2-4-7(5)9/h2-4H,9H2,1H3

InChI key

SLDLVGFPFFLYBM-UHFFFAOYSA-N

application

3-Fluoro-2-methylaniline was used in the synthesis of 2-amino-1,7,9-trimethylimidazo[4,5-g]quinoxaline[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

186.8 °F - closed cup

flash_point_c

86 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Achiwa et al.
Chemical & pharmaceutical bulletin, 42(2), 408-409 (1994-02-01)
A new mutagen, 2-amino-1,7,9-trimethylimidazo[4,5-g]quinoxaline (1), isolated from beef extract, was synthesized from 3-fluoro-2-methylaniline via an intermediate, 2,8-dimethyl-7-methylaminoquinoxaline (7a). Its 2-methylanalog (2) was also synthesized from the same intermediate. The synthetic 1 showed the same mutagenic activity as the isolated mutagen.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service