Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217751

Sigma-Aldrich

4-Fluoro-2-(trifluoromethyl)aniline

99%

Synonym(s):

2-Amino-5-fluorobenzotrifluoride, α,α,α,4-Tetrafluoro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CF3)NH2
CAS Number:
Molecular Weight:
179.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

70-72 °C/17.5 mmHg (lit.)

density

1.38 g/mL at 25 °C (lit.)

functional group

fluoro

storage temp.

2-8°C

SMILES string

Nc1ccc(F)cc1C(F)(F)F

InChI

1S/C7H5F4N/c8-4-1-2-6(12)5(3-4)7(9,10)11/h1-3H,12H2

InChI key

LRCQLCWUUBSUOY-UHFFFAOYSA-N

Application

4-Fluoro-2-(trifluoromethyl)aniline was used in the preparation of 2-[4-(3-bromophenyl)-7-chloro-6-methyl-2-oxo-2H-chromen-3-yl]-N-[4-fluoro-2-(trifluoro-methyl)phenyl]acetamide[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaki Ogino et al.
Chemical & pharmaceutical bulletin, 59(11), 1369-1375 (2011-11-02)
As a part of our research for novel potent and orally available acyl-CoA: cholesterol acyltransferase (ACAT) inhibitors that can be used as anti-atherosclerotic agents, we recently reported the discovery of the (4-phenylcoumarine)acetanilide derivative 1. However, compound 1 showed adrenal toxicity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service