Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

213012

Sigma-Aldrich

Benzeneseleninic acid anhydride

70%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SeOOSeOC6H5
CAS Number:
Molecular Weight:
360.13
Beilstein/REAXYS Number:
2332406
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

70%

form

powder

impurities

<30% benzeneseleninic acid

mp

165-170 °C (lit.)

SMILES string

O=[Se](O[Se](=O)c1ccccc1)c2ccccc2

InChI

1S/C12H10O3Se2/c13-16(11-7-3-1-4-8-11)15-17(14)12-9-5-2-6-10-12/h1-10H

InChI key

FHPZOWOEILXXBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxidation of phenols by benzeneseleninic acid anhydride yields ortho-quinones[1]. Benzeneseleninic anhydride acts as a mild oxidant for the conversion of benzylic hydrocarbons into aldehydes or ketones[2].

Application

Benzeneseleninic acid anhydride was used to oxidize hydrazines to afford azo-compounds[3].

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydrogenation of hydrazines and of 4-azacholestan-3-one with benzeneseleninic acid and benzeneseleninic anhydride.
Back TG.
Journal of the Chemical Society. Chemical Communications, 6, 278-279 (1978)
Comparative oxidation of phenols with benzeneseleninic anhydride and with benzeneseleninic acid.
Barton DHR, et al.
Tetrahedron, 44(20), 6397-6406 (1988)
Preparation of aldehydes and ketones by oxidation of benzylic hydrocarbons with benzeneseleninic anhydride.
Barton DHR, et al.
Tetrahedron Letters, 20(35), 3331-3334 (1979)
Naotoshi Toki et al.
Chemical & pharmaceutical bulletin, 52(8), 1009-1012 (2004-08-12)
Benzeneseleninic anhydride in the presence of tert-butyl hydroperoxide in chlorobenzene at about 70 degrees C is an effective oxidizing agent for the selective oxidation of alcohols at the benzylic position.
T Iida et al.
Journal of lipid research, 29(8), 1097-1101 (1988-08-01)
A facile one-step conversion of stereoisomeric methyl 3-hydroxycholanoates to 1,4-dien-3-one, by treatment in boiling toluene with iodoxybenzene catalyzed by benzeneselenic anhydride, is described. The direct oxidation-dehydrogenation at C-3 is applicable to other cholanoates (hyodeoxycholic, chenodeoxycholic, ursodeoxycholic, deoxycholic, and cholic) when

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service