All Photos(1)
About This Item
Linear Formula:
Rh2O3 · xH2O
CAS Number:
Molecular Weight:
253.81 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form
powder
composition
Rh, 55%
reaction suitability
reagent type: catalyst
core: rhodium
SMILES string
O.O=[Rh]O[Rh]=O
InChI
1S/H2O.3O.2Rh/h1H2;;;;;
InChI key
XLXOEVBEAKJARU-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service