Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

194034

Sigma-Aldrich

N-Methyl-2-nitroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NHCH3
CAS Number:
Molecular Weight:
152.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

35-38 °C (lit.)

functional group

amine
nitro

SMILES string

CNc1ccccc1[N+]([O-])=O

InChI

1S/C7H8N2O2/c1-8-6-4-2-3-5-7(6)9(10)11/h2-5,8H,1H3

InChI key

KFBOUJZFFJDYTA-UHFFFAOYSA-N

General description

N-Methyl-2-nitroaniline is a solvatochromic dye[1].

Application

N-Methyl-2-nitroaniline was used in the determination of the polarities of ionic liquids using the Kamlet-Taft empirical polarity scales[1]. It was also employed in characterization of the octadecyl-derivatized silica stationary phase and the corresponding mobile phases used in reversed-phase liquid chromatography[2].

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvatochromic studies on reversed-phase liquid chromatographic phases. 2. Characterization of stationary and mobile phases.
Lu H and Rutan SC.
Analytical Chemistry, 68(8), 1387-1393 (1996)
M A Ab Rani et al.
Physical chemistry chemical physics : PCCP, 13(37), 16831-16840 (2011-08-23)
The polarities of a wide range of ionic liquids have been determined using the Kamlet-Taft empirical polarity scales α, β and π*, with the dye set Reichardt's Dye, N,N-diethyl-4-nitroaniline and 4-nitroaniline. These have been compared to measurements of these parameters
Dilek Yalcin et al.
Physical chemistry chemical physics : PCCP, 22(1), 114-128 (2019-12-10)
Ionic liquids (ILs) are highly tailorable solvents with many potential applications. Knowledge about their solvation properties is highly beneficial in the utilization of ILs for specific tasks, though for many ILs this is currently unknown. In this study, we have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service