Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

193461

2-Ethoxyethyl isobutyrate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O3
CAS Number:
Molecular Weight:
160.21
PubChem Substance ID:

assay

≥99%

InChI

1S/C8H16O3/c1-4-10-5-6-11-8(9)7(2)3/h7H,4-6H2,1-3H3

InChI key

ANSOCBLHUNAGRK-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service