Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

191388

Sigma-Aldrich

2-Ethoxybenzylamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4CH2NH2
CAS Number:
Molecular Weight:
151.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

refractive index

n20/D 1.533 (lit.)

bp

69-74 °C/0.2 mmHg (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

CCOc1ccccc1CN

InChI

1S/C9H13NO/c1-2-11-9-6-4-3-5-8(9)7-10/h3-6H,2,7,10H2,1H3

InChI key

LAUPTNYHVCVPFH-UHFFFAOYSA-N

General description

2-Ethoxybenzylamine has been used in the preparation of 2,3,6-trisubstituted pyridines[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Lucchesini et al.
Bioorganic & medicinal chemistry, 22(5), 1558-1567 (2014-02-18)
In order to obtain substrate-like inhibitors of copper amine oxidases (CAOs), a class of enzymes involved in important cellular processes as well as in crosslinking of elastin and collagen and removal of biogenic primary amines, we synthesized a set of
The Bohlmann-Rahtz route to functionalised pyridine scaffolds and their use in library synthesis.
Bashford KE, et al.
Tetrahedron Letters, 44(8), 1627-1629 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service