187275
3-Cyclopentyl-1-propanol
99%
Synonym(s):
Cyclopentanepropanol
About This Item
assay
99%
refractive index
n20/D 1.459 (lit.)
bp
93-95 °C/8 mmHg (lit.)
density
0.907 g/mL at 25 °C (lit.)
SMILES string
OCCCC1CCCC1
InChI
1S/C8H16O/c9-7-3-6-8-4-1-2-5-8/h8-9H,1-7H2
InChI key
IBMXMCXCSPGCDQ-UHFFFAOYSA-N
application
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
179.6 °F - closed cup
flash_point_c
82 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service