Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

179531

Sigma-Aldrich

Butyl levulinate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2CO2(CH2)3CH3
CAS Number:
Molecular Weight:
172.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

98%

form

liquid

refractive index

n20/D 1.427 (lit.)

bp

106-108 °C/5.5 mmHg (lit.)

density

0.974 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)CCC(C)=O

InChI

1S/C9H16O3/c1-3-4-7-12-9(11)6-5-8(2)10/h3-7H2,1-2H3

InChI key

ISBWNEKJSSLXOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Butyl levulinate was used in the synthesis of γ-valerolactone[1].

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xian-Long Du et al.
ChemSusChem, 4(12), 1838-1843 (2011-11-23)
The utilization of biomass has recently attracted tremendous attention as a potential alternative to petroleum for the production of liquid fuels and chemicals. We report an efficient alcohol-mediated reactive extraction strategy by which a hydrophobic mixture of butyl levulinate and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service