Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

177555

Sigma-Aldrich

2,5-Dichloroiodobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3I
CAS Number:
Molecular Weight:
272.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.646 (lit.)

bp

255-256 °C/742 mmHg (lit.)

mp

21 °C (lit.)

density

2.026 g/mL at 25 °C (lit.)

functional group

chloro
iodo

SMILES string

Clc1ccc(Cl)c(I)c1

InChI

1S/C6H3Cl2I/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

SBHVNORGKIPGCL-UHFFFAOYSA-N

Application

2,5-Dichloroiodobenzene may be used in the synthesis of thioether bond-containing aromatic diamines.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

201.2 °F - closed cup

flash_point_c

94 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service