Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

176451

Sigma-Aldrich

γ-Phenyl-γ-butyrolactone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

solid

bp

306 °C (lit.)

mp

36-37 °C (lit.)

density

1.155 g/mL at 25 °C (lit.)

SMILES string

O=C1CCC(O1)c2ccccc2

InChI

1S/C10H10O2/c11-10-7-6-9(12-10)8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

AEUULUMEYIPECD-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R O Juvonen et al.
Xenobiotica; the fate of foreign compounds in biological systems, 30(1), 81-92 (2000-02-05)
1. The structural requirements for a compound to be a potent inhibitor for mouse CYP2A5 and human CYP2A6 enzymes catalysing coumarin 7-hydroxylase activity have been studied. 2. The IC50 of 28 compounds for the pyrazole-treated male DBA/2 mouse and human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service