Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

168114

Sigma-Aldrich

4-tert-Butylbenzoic hydrazide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4CONHNH2
CAS Number:
Molecular Weight:
192.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

120-127 °C (lit.)

functional group

amine
hydrazine

SMILES string

CC(C)(C)c1ccc(cc1)C(=O)NN

InChI

1S/C11H16N2O/c1-11(2,3)9-6-4-8(5-7-9)10(14)13-12/h4-7H,12H2,1-3H3,(H,13,14)

InChI key

XYUFQWDLRLHUPB-UHFFFAOYSA-N

Application

4-tert-Butylbenzoic hydrazide was used in the synthesis of 2-(2-methylphenyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saghir Abbas et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 15(3), 298-310 (2018-10-17)
The over-expression of the carbonic anhydrases results in some specific carcinomas including pancreatic, gastric and brain tumor. Tumors are distinguished under hypoxic conditions and various investigations are being carried out to target the known hypoxic areas of the tumors to
Zn2+ selective luminescent 'off-on'probes derived from diaryl oxadiazole and aza-15-crown-5.
Mashraqui SH, et al.
Tetrahedron, 63(45), 11093-11100 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service