Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

165018

Sigma-Aldrich

Methyl 5-chlorosalicylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$252.90

$252.90

List Price$281.00Save 10%

Estimated to ship onApril 26, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$252.90

About This Item

Empirical Formula (Hill Notation):
C8H7ClO3
CAS Number:
Molecular Weight:
186.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$252.90

List Price$281.00Save 10%

Estimated to ship onApril 26, 2025


Request a Bulk Order

Quality Level

assay

97%

mp

46-50 °C (lit.)

functional group

chloro
ester

SMILES string

COC(=O)c1cc(Cl)ccc1O

InChI

1S/C8H7ClO3/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4,10H,1H3

InChI key

KJWHRMZKJXOWFC-UHFFFAOYSA-N

General description

Methyl 5-chlorosalicylate on iodination yields 3-iodo-5-chlorosalicylate[1].

Application

Methyl 5-chlorosalicylate was used in the synthesis of N-p-methoxyphenylbenzimino-p-chloro-o-carbomethoxyphenyl ether[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

411. Some derivatives of diphenylamine and a new synthesis of N-arylanthranilic acids and of acridones.
Jamison MM and Turner EE.
Journal of the Chemical Society, 1954-1959 (1937)
Halogenosalicylohydroxamic Acids. I. Dihalogenosalicylohydroxamic Acids.
DUDA H, et al.
Bulletin de l'Academie Polonaise des Sciences, Serie des Sciences Chimiques, 13(5), 341-347 (1965)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service