Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

163953

Sigma-Aldrich

3-Benzyloxy-4-methoxybenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
242.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

98%

form

solid

mp

61-64 °C (lit.)

functional group

aldehyde
phenyl

SMILES string

COc1ccc(C=O)cc1OCc2ccccc2

InChI

1S/C15H14O3/c1-17-14-8-7-13(10-16)9-15(14)18-11-12-5-3-2-4-6-12/h2-10H,11H2,1H3

InChI key

VQVQZFHUXRSRBZ-UHFFFAOYSA-N

Application

3-Benzyloxy-4-methoxybenzaldehyde was used in the synthesis of (+)-9-benzyloxy-α-dihydrotetrabenazine[1]. It was also used in total synthesis of (-)-galipeine[2].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyi Liu et al.
Chirality, 25(4), 215-223 (2013-03-28)
This article reports, for the first time, on the absolute configuration of (+)-9-benzyloxy-α-dihydrotetrabenazine (8), as determined from the perspective of X-ray crystallography. Compound 8 was prepared by a six-step reaction using 3-benzyloxy-4-methoxybenzaldehyde (1) as a starting material. The X-ray crystal
The enantioselective total synthesis of alkaloid (-)-galipeine.
Yang P-Y and Zhou Y-G.
Tetrahedron Asymmetry, 15(7), 1145-1149 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service