Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

163368

Sigma-Aldrich

7-Methoxy-1-tetralone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
Beilstein/REAXYS Number:
2208389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

solid

mp

59-63 °C (lit.)

functional group

ketone

SMILES string

COc1ccc2CCCC(=O)c2c1

InChI

1S/C11H12O2/c1-13-9-6-5-8-3-2-4-11(12)10(8)7-9/h5-7H,2-4H2,1H3

InChI key

GABLTKRIYDNDIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

7-methoxy-1-tetralone was used in the synthesis of 2,7-dimethoxy-1-tetralone[1], substituted derivatives of 3a,4,5,6-tetrahydrosuccinimido[3,4-b]acenaphthen-10-one[2] and 13H-benzo[6-7]indolo[3,2-c]quinolines[3].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

13H-benzo [6-7] indolo [3, 2-c] quinolines (B [6, 7] IQ): optimization of their DNA triplex-specific stabilization properties.
Schmitt P, et al.
Chemical Communications (Cambridge, England), 9, 763-764 (2000)
Substituted Derivatives of 3a, 4, 5, 6-Tetrahydrosuccinimido [3, 4-b] Acenaphthen-10-one.
Campaigne E and Weddleton RF.
Proceedings of the Indian Academy of Sciences - Section A, 95 (2013)
The reaction of 1-tetralones with thallium trinitrate supported on clay: ring contraction vs alpha-oxidation.
Ferraz H, et al.
Journal of the Brazilian Chemical Society, 12(5), 680-684 (2001)
Tian-Min Wang et al.
Journal of separation science, 40(17), 3440-3452 (2017-07-07)
As a traditional medicinal plant, Juglans mandshurica has been used for the treatment of cancer. Different organs of this plant showed anti-tumor activity in clinic and laboratory. Comparative identification of constituents in different plant organs is essential for investigation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service