Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

159093

Sigma-Aldrich

2,6-Dinitroaniline

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3NH2
CAS Number:
Molecular Weight:
183.12
Beilstein/REAXYS Number:
2214886
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

134 °C (dec.) (lit.)

functional group

nitro

SMILES string

Nc1c(cccc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H5N3O4/c7-6-4(8(10)11)2-1-3-5(6)9(12)13/h1-3H,7H2

InChI key

QFUSCYRJMXLNRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dinitroaniline is a dinitroaniline. It serves as precursor to various new 1,2,3-trisubstituted halobenzene derivatives.[1]

Application

2,6-Dinitroaniline was used in the preparation of 2,6-dinitroiodobenzene.[1]

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

>212.0 °F

flash_point_c

> 100 °C

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and NMR analysis of 2, 6-heterodifunctional halobenzenes as precursors for substituted biphenyls
Sienkowska M, et al.
Tetrahedron, 56(2), 165-173 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service