Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

155683

Sigma-Aldrich

3-Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-oxyl

99%

Synonym(s):

2,2,5,5-Tetramethyl-3-carbamido-3-pyrroline-1-oxyl, 3-Carbamoyl-2,5-dihydro-2,2,5,5-tetramethyl-1H-pyrrol-1-yloxy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N2O2
CAS Number:
Molecular Weight:
183.23
Beilstein/REAXYS Number:
4138020
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

solid

mp

197-200 °C (lit.)

functional group

amide

storage temp.

2-8°C

SMILES string

CC1(C)C=C(C(N)=O)C(C)(C)N1[O]

InChI

1S/C9H15N2O2/c1-8(2)5-6(7(10)12)9(3,4)11(8)13/h5H,1-4H3,(H2,10,12)

InChI key

RUEXQFPLRRIFTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-oxyl is a nitroxide radical that can be used:
  • As a spin probe in the study of simulation of Overhauser dynamic nuclear polarization signal.[1]
  • As a starting material in the synthesis of nitroxide based polyethers possessing charge transport properties.[2]
  • As a nitroxide imaging probe applicable for EPR imaging of brain diseases in animal models.[3]

Spin label

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B H Robinson et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 138(2), 210-219 (1999-05-26)
This work demonstrates that homogeneous linewidths can be extracted from continuous wave electron paramagnetic resonance spectra and that they quantitatively agree with the predictions of existing relaxation theory. We suggest that relaxation theory can be used to predict experimental lineshapes
Ichiro Koshiishi et al.
The Biochemical journal, 395(2), 303-309 (2006-01-07)
Lipoxygenases have been proposed to be a possible factor that is responsible for the pathology of certain diseases, including ischaemic injury. In the peroxidation process of linoleic acid by lipoxygenase, the E,Z-linoleate allyl radical-lipoxygenase complex seems to be generated as
F Gomi et al.
Life sciences, 52(25), 2027-2033 (1993-01-01)
After the nitroxyl radical 3-carbamoyl-2,2,5,5-tetramethyl-1-pyrrolidinyloxyl (carbamoylPROXYL) was intraperitoneally administered to mice, the time courses of changes in the intensity of signals in L-band electron spin resonance spectra from their brains were measured. The spin clearance rate of old, ad libitum-fed
S Cavalu et al.
Biomacromolecules, 4(6), 1630-1635 (2003-11-11)
Noncovalent spin labeled proteins (ovalbumin, bovine serum albumin, hemoglobin, and cytochrome c) were investigated in order to follow the different type of interactions between the nitroxide radical of 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy spin label and functional groups of heme and nonheme proteins as
H J Halpern et al.
Radiation research, 145(5), 610-618 (1996-05-01)
We have used very low-frequency electron paramagnetic resonance (EPR) oximetry to measure the change in oxygen concentration (delta pO2) due to change in breathing atmosphere in FSa and NFSa fibrosarcomas implanted in the legs of C3H mice infused with perfluoro-octylbromine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service