Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

155667

Sigma-Aldrich

2-Chlorothiophene

96%

Synonym(s):

2-Thienyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3ClS
CAS Number:
Molecular Weight:
118.58
Beilstein/REAXYS Number:
104652
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

liquid

refractive index

n20/D 1.547 (lit.)

bp

127-129 °C (lit.)

density

1.286 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1cccs1

InChI

1S/C4H3ClS/c5-4-2-1-3-6-4/h1-3H

InChI key

GSFNQBFZFXUTBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The photodissociation dynamics of 2-chlorothiophene was studied using resonance enhanced multiphoton ionization (REMPI) time-of-flight (TOF) technique[1].

Application

2-Chlorothiophene was used in the preparation of 5-phenylthiophene derivative[2].

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

73.4 °F - closed cup

flash_point_c

23 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Steinmetz et al.
Chemistry, an Asian journal, 7(6), 1256-1260 (2012-02-16)
Direct C-H phenylation of 2-ethylthiophene and 2-chlorothiophene with PhPdI(bipy) complex to form either the corresponding 4-phenyl or 5-phenylthiophene derivative is studied under stoichiometric conditions using various Lewis acids as additives. It is shown that reactions occur via the corresponding cationic
Monali Kawade et al.
The journal of physical chemistry. A, 116(44), 10656-10667 (2012-10-20)
The photodissociation dynamics of halogen-substituted thiophenes, namely, 2-chlorothiophene and 2-bromo-5-chlorothiophene, has been studied in a supersonic molecular beam around 235 nm, using resonance enhanced multiphoton ionization (REMPI) time-of-flight (TOF) technique, by detecting the nascent state of the primary halogen atoms.
Hari P Upadhyaya
Rapid communications in mass spectrometry : RCM, 33(20), 1598-1612 (2019-05-31)
Halogenated thiophenes are an important class of compounds mostly used in the synthesis of various materials, showing unusual electronic and optical properties. The Thiophene Ring Fragmentation (TRF) process is widely used in synthetic chemistry. In this study, the fragmentation pattern

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service