Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15385

Sigma-Aldrich

Boc-Ala-OSu

Synonym(s):

Boc-L-alanine N-succinimidyl ester, Boc-L-alanine hydroxysuccinimide ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O6
CAS Number:
Molecular Weight:
286.28
Beilstein/REAXYS Number:
3560476
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]20/D −52±3.5°, c = 2.5% in dioxane

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

161-165 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

InChI

1S/C12H18N2O6/c1-7(13-11(18)19-12(2,3)4)10(17)20-14-8(15)5-6-9(14)16/h7H,5-6H2,1-4H3,(H,13,18)/t7-/m0/s1

InChI key

COMUWNFVTWKSDT-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in synthesis of:
  • Muramyldipeptide analogs via solid-phase synthesis
  • Microcin C analogs for use as antibacterials
  • Lipid I and nucleoside diphosphate peptide derivatives via N-methylimidazolium chloride catalyzed coupling
  • N-Protected dipeptide acids
  • Free and peptide-bound glycated amino acids for studies of interactions with human Caco-2 interstinal epithelial cell apical membrane transport proteins
  • 7-Substituted camptothecin conjugates with RGD-peptides as avβ3 integrin ligands

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service