Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

147990

Supelco

(R)-(−)-2-Octanol

derivatization grade (chiral), LiChropur, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
Beilstein/REAXYS Number:
1719324
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.05

grade

derivatization grade (chiral)

assay

99%

form

liquid

optical activity

[α]17/D −9.5°, neat

quality

LiChropur

technique(s)

HPLC: suitable

refractive index

n20/D 1.426 (lit.)

bp

175 °C (lit.)

density

0.820 g/mL at 20 °C (lit.)

SMILES string

CCCCCC[C@@H](C)O

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3/t8-/m1/s1

InChI key

SJWFXCIHNDVPSH-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(−)-2-Octanol is a chiral derivatizating agent, which is employed for derivatizing enantiomers into diastereoisomers.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

168.8 °F - closed cup

flash_point_c

76 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timofey V Malyarenko et al.
Molecules (Basel, Switzerland), 23(5) (2018-05-13)
Two new polyhydroxysteroidal glycosides, anthenosides A₁ (1) and A₂ (2), and one previously known steroidal glycoside anthenoside A (3) were isolated from extract of the tropical starfish Anthenea aspera. Structures of 1⁻3 were determined by analysis of the spectroscopic data
Paul Masset et al.
Cell, 182(1), 112-126 (2020-06-07)
Every decision we make is accompanied by a sense of confidence about its likely outcome. This sense informs subsequent behavior, such as investing more-whether time, effort, or money-when reward is more certain. A neural representation of confidence should originate from
Xiao-Yang Ou et al.
Journal of biotechnology, 299, 37-43 (2019-05-03)
Highly efficient asymmetric reduction of 2-octanone to (R)-2-octanol catalyzed by immobilized Acetobacter sp. CCTCC M209061 cells was achieved in a biphasic system. Bioreduction conducted in aqueous single phase buffer was limited due to poor solubility and toxicity towards cells cause
Anindya S Bhattacharjee et al.
Cell reports, 28(11), 2966-2978 (2019-09-12)
The olfactory environment is first represented by glomerular activity patterns in the olfactory bulb. It remains unclear how these representations intersect with sampling behavior to account for the time required to discriminate odors. Using different chemical classes, we investigate glomerular
Deracemization of secondary alcohols through a concurrent tandem biocatalytic oxidation and reduction.
Constance V Voss et al.
Angewandte Chemie (International ed. in English), 47(4), 741-745 (2007-12-12)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service