Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

144177

Sigma-Aldrich

4-Methoxycinnamonitrile, mixture of cis and trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH=CHCN
CAS Number:
Molecular Weight:
159.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.6131 (lit.)

bp

127-140 °C/1 mmHg (lit.)

density

1.096 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(\C=C\C#N)cc1

InChI

1S/C10H9NO/c1-12-10-6-4-9(5-7-10)3-2-8-11/h2-7H,1H3/b3-2+

InChI key

CNBCNHHPIBKYBO-NSCUHMNNSA-N

General description

4-Methoxycinnamonitrile shows potent nematicidal activity toward adult Bursaphelenchus xylophilus[1]. Surface-enhanced Raman scattering of cis- and trans-4-methoxycinnamonitrile has been studied in aqueous silver sols[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong-Ok Kong et al.
Journal of nematology, 39(1), 31-36 (2007-03-01)
The nematicidal activity of two cassia, Cinnamomum cassia, oils (Especial and true), four cinnamon, Cinnamomum zey-lanicum, oils (technical, #500, bark and green leaf), and their compounds (e.g., trans-cinnamaldehyde and trans-cinnamic acid) toward adult Bursaphelenchus xylophilus was examined by a direct
Adsorption and reaction of 4-methoxycinnamonitrile on a silver surface: A surface-enhanced Raman spectroscopic study.
Chun HA, et al.
Journal of Raman Spectroscopy, 21(11), 743-749 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service