Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

123625

Sigma-Aldrich

2-Benzoylacetanilide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2CONHC6H5
CAS Number:
Molecular Weight:
239.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

106-108 °C (lit.)

solubility

acetone: soluble 25 mg/mL, clear, colorless to faintly yellow

SMILES string

O=C(CC(=O)c1ccccc1)Nc2ccccc2

InChI

1S/C15H13NO2/c17-14(12-7-3-1-4-8-12)11-15(18)16-13-9-5-2-6-10-13/h1-10H,11H2,(H,16,18)

InChI key

XRZDIHADHZSFBB-UHFFFAOYSA-N

General description

2-Benzoylacetanilide reacts with [PtCl2(cod)] (cod=cyclo-octa-1,5-diene), cis-[PtCl2(PPh3)2] (PPh3= triphenylphosphine) and [PdCl2(bipy)] (bipy=2,2′-bipyridine) to yield metallalactam complexes[1].

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Platina-and palladalactam complexes derived from 2-benzoylacetanilide; syntheses and X-ray structure of [Pd {NPhC (O) CHC (O) Ph}(bipy)]? CH2Cl2.
Henderson W, et al.
Inorgorganica Chimica Acta, 292(2), 260-265 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service