Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

119148

Sigma-Aldrich

exo-6-Hydroxytropinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

mp

120-121 °C (lit.)

SMILES string

CN1[C@@H]2C[C@H](O)[C@H]1CC(=O)C2

InChI

1S/C8H13NO2/c1-9-5-2-6(10)4-7(9)8(11)3-5/h5,7-8,11H,2-4H2,1H3/t5-,7+,8-/m0/s1

InChI key

UOHSTKWPZWFYTF-ARDNSNSESA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Hydroxytropinone was used in the synthesis of N-substituted 6-ethyl or -methyl-8-hydroxy-1,2,3,4,5,6-hexahydro-1,4:2,6-dimethano-3-benzazocines[1].

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Montzka et al.
Journal of medicinal chemistry, 20(3), 453-456 (1977-03-01)
A seres of (+/-)-N-substituted 6-ethyl-or -methyl-8-hydroxy-1,2,3,4,5,6-hexahydro-1,4:2,6-dimethano-3-benzazocines has been prepared from 6 hydroxytropinone. The N-cyclopropylmethyl compounds 10a and 10b were found to be strong narcotic antagonists approximately equivalent to nalorphine. Only slight analgetic activity was found in any of these compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service