Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

113514

Sigma-Aldrich

2-Amino-3-nitropyridine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N3O2
CAS Number:
Molecular Weight:
139.11
Beilstein/REAXYS Number:
124468
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

solid

mp

163-165 °C (lit.)

functional group

nitro

SMILES string

Nc1ncccc1[N+]([O-])=O

InChI

1S/C5H5N3O2/c6-5-4(8(9)10)2-1-3-7-5/h1-3H,(H2,6,7)

InChI key

BPYHGTCRXDWOIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Amino-3-nitropyridine functionalizes Wang resin via a carbamate linkage.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

332.6 °F - closed cup

flash_point_c

167 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Ermann et al.
Journal of combinatorial chemistry, 4(4), 352-358 (2002-07-09)
A solid-phase synthesis of substituted cyclic urea derivatives as potential heterocyclic library scaffolds is described. 2-Amino-3-nitropyridine is attached to Wang resin via a carbamate linkage. Reduction of the nitro group was achieved with SnCl(2).2H(2)O. Reductive alkylation with a range of
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service