Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900936

Nystatin

Vetec, reagent grade, ≥4400 USP units/mg

Synonym(s):

Nystatin, Fungicidin, Mycostatin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

reagent grade

product line

Vetec

concentration

≥4400 USP units/mg

storage temp.

−20°C

SMILES string

O[C@@H]([C@H](C)[C@H](C)O1)[C@@H](C)/C=C/C=C/CC/C=C/C=C/C=C/C=C/[C@H](O[C@@H]2O[C@H](C)[C@@H](O)[C@H](N)[C@@H]2O)C[C@@]3([H])[C@H](C(O)=O)[C@@H](O)C[C@](O)(O3)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)C[C@@H](O)CC1=O

InChI

1S/C47H75NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-36(53)35(52)20-19-31(49)21-32(50)22-33(51)23-39(55)62-29(3)28(2)42(27)56/h5-6,8,10-18,27-38,40-44,46,49-54,56-58,61H,7,9,19-26,48H2,1-4H3,(H,59,60)/b6-5+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28+,29-,30+,31+,32+,33+,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

VQOXZBDYSJBXMA-QEKUPDCNSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mode of Action: Nystatin acts by binding to ergosterols in fungal cell membranes, increasing permeability of the membrane, and creating nystatin/ergosterol based ion channels. It also functions as a lipid raft-inhibiting reagent and as membrane associated cholesterol.

Antimicrobial spectrum: Nystatin acts against fungi, yeasts and molds.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service