Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900934

Alizarin Red S

Vetec, reagent grade

Synonym(s):

Alizarin Red S, 3,4-Dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt, Alizarin Carmine, Alizarin Red, water soluble, Alizarin sodium monosulfonate, Alizarin sulfonate sodium, Alizarinsulfonic acid sodium salt, Sodium alizarinsulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7NaO7S
CAS Number:
Molecular Weight:
342.26
Colour Index Number:
58005
Beilstein/REAXYS Number:
4117090
MDL number:
UNSPSC Code:
41116004
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Oc1c(O)c(cc2C(=O)c3ccccc3C(=O)c12)S([O-])(=O)=O

InChI

1S/C14H8O7S.Na/c15-11-6-3-1-2-4-7(6)12(16)10-8(11)5-9(22(19,20)21)13(17)14(10)18;/h1-5,17-18H,(H,19,20,21);/q;+1/p-1

InChI key

HFVAFDPGUJEFBQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sigeng Chen et al.
Bone, 67, 109-121 (2014-07-08)
Although osteoinductive ceramics can induce osteoblast differentiation in vitro and bone regeneration in vivo, their effects rely solely on the limited number of endogenous stem cells. More recently, ceramic carriers seeded with culture-expanded stem cells have been reported as implants

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service