V900914
DirectLoad™ 1 kb DNA Ladder
Vetec™, reagent grade, ready-to-use marker for DNA electrophoresis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
reagent grade
product line
Vetec™
form
liquid
usage
100 uses
suitability
suitable for electrophoresis (DNA)
Looking for similar products? Visit Product Comparison Guide
General description
Vetec 1 kb Ladder contains 12 fragments consisting of 300 bp, 500 bp, 800 bp, 1 kb, 1.5 kb, 2 kb, 3 kb, 4 kb, 5 kb, 6 kb, 8 kb, 10 kb bands. Five μl of the ladder can be loaded directly into a single lane on an agarose (V900510) or polyacrylamide gel.
Application
Suitable for size determination of dsDNA by DNA electrophoresis with either agarose or polyacrylamide gels.
Components
Vetec 1kb DNA Ladder is provided in a gel-loading solution of 10 mM Tris-HCl, 10 mM EDTA, 0.02% bromophenol blue, and 5% glycerol.
Other Notes
Contains 12 fragments consisting of 300 bp, 500 bp, 800 bp, 1 kb, 1.5 kb, 2 kb, 3 kb, 4 kb, 5 kb, 6 kb, 8 kb, 10 kb bands.
For optimal resolution, the recommended agarose gel concentration is 0.8-1.0%.
Legal Information
DirectLoad is a trademark of Sigma-Aldrich Co. LLC
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service