Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900894

Sigma-Aldrich

Paraformaldehyde

Vetec, reagent grade, powder

Synonym(s):

Polyoxymethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2O)nH
CAS Number:
Molecular Weight:
30.03 (as monomer)
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

grade

reagent grade

vapor density

1.03 (vs air)

vapor pressure

<1.45 mmHg ( 25 °C)

product line

Vetec

form

powder

autoignition temp.

572 °F

expl. lim.

73 %

mp

120-170 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

storage temp.

2-8°C

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zhengyu Luo et al.
iScience, 23(4), 101034-101034 (2020-04-22)
Three-dimensional chromatin structures undergo dynamic reorganization during mammalian spermatogenesis; however, their impacts on gene regulation remain unclear. Here, we focused on understanding the structure-function regulation of meiotic chromosomes by Hi-C and other omics techniques in mouse spermatogenesis across five stages.
Assessment of cytokines by immunofluorescence and the paraformaldehyde-saponin procedure.
B Sander et al.
Immunological reviews, 119, 65-93 (1991-02-01)
Zhen Yang et al.
Autophagy, 15(4), 668-685 (2018-11-06)
Emerging evidence has revealed that miRNAs could upregulate the expression levels of target genes. However, the molecular mechanism underlying upregulation of targets mediated by miRNAs remains unclear. In this study, we found a novel miRNA named MIR-G-1 by GRSF1-RNA immunoprecipitation
Ye Jee Shim et al.
Laboratory medicine, 45(2), 120-131 (2014-05-30)
The granulocyte immunofluorescence test based on flow cytometry is still the most commonly used method for detecting antineutrophil antibodies. We questioned whether neutrophil prefixation is necessary for this test. Three pretreatment methods using paraformaldehyde were compared: unfixed, short-fixed (3 minutes
N Kotova et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(7), 1897-1905 (2015-07-25)
The induction of micronucleated reticulocytes in the bone marrow is a sensitive indicator of chromosomal damage. Therefore, the micronucleus assay in rodents is widely used in genotoxicity and carcinogenicity testing. A test system based on cultured human primary cells could

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service