Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900851

Sigma-Aldrich

SSPE Buffer 20× Concentrate

Vetec, reagent grade, 20 ×, solution, RNase and DNase free

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319

grade

reagent grade

product line

Vetec

form

solution

feature

RNase and DNase free

concentration

20 ×

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service