Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900834

Sigma-Aldrich

Reserpine

Vetec, reagent grade

Synonym(s):

(3β, 16β, 17α, 18β, 20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40N2O9
CAS Number:
Molecular Weight:
608.68
Beilstein/REAXYS Number:
102014
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

mp

~265 °C (dec.)

SMILES string

CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)c6cc(OC)c(OC)c(OC)c6

InChI

1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1

InChI key

QEVHRUUCFGRFIF-MDEJGZGSSA-N

Gene Information

human ... SLC18A2(6571)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits vesicular uptake of catecholamines and serotonin.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1A - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reserpine: a challenge for total synthesis of natural products.
Fen-Er Chen et al.
Chemical reviews, 105(12), 4671-4706 (2005-12-15)
Sandy D Shamon et al.
The Cochrane database of systematic reviews, (4)(4), CD007655-CD007655 (2009-10-13)
Many antihypertensive agents exist today for the treatment of primary hypertension (systolic blood pressure >/=140 mmHg and/or diastolic blood pressure >/=90 mmHg). Randomised controlled trials have been carried out to investigate the evidence for these agents.There is, for example, strong
Reserpine-induced supersensitivity to the cardiac effects of agonists.
K D Meisheri et al.
Life sciences, 24(6), 473-480 (1979-02-05)
How reserpine and chlorpromazine act: the impact of key discoveries on the history of psychopharmacology.
G Curzon
Trends in pharmacological sciences, 11(2), 61-63 (1990-02-01)
R E Stitzel
Pharmacological reviews, 28(3), 179-208 (1976-09-01)
Orally administered reserpine is readily absorbed from the GI tract. During this process at least a portion of the drug is metabolized by the intestinal mucosa and then presumably is acted upon by serum esterases. Methylreserpate and trimethoxybenzoic acid are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service