Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900832

Sigma-Aldrich

Phosphocreatine disodium salt hydrate

98%, Vetec

Synonym(s):

N-(Imino[phosphonoamino]methyl)-N-methylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H8N3O5PNa2 · x H2O
CAS Number:
Molecular Weight:
255.08 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

Product Name

Phosphocreatine disodium salt hydrate, Vetec, reagent grade, 98%

grade

reagent grade

product line

Vetec

assay

98%

form

powder

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].CN(CC(O)=O)C(=N)NP([O-])([O-])=O

InChI

1S/C4H10N3O5P.2Na.H2O/c1-7(2-3(8)9)4(5)6-13(10,11)12;;;/h2H2,1H3,(H,8,9)(H4,5,6,10,11,12);;;1H2/q;2*+1;/p-2

InChI key

KZVYPOKBYRXZKS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phosphocreatine, a high-energy phosphate reservoir in vertebrate and some invertebrate muscles, provides phosphate for ADP-ATP conversion.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service