Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900829

Sigma-Aldrich

Palladium(II) chloride

Vetec, reagent grade, 98%

Synonym(s):

Dichloropalladium, Palladium dichloride, Palladous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Pd
CAS Number:
Molecular Weight:
177.33
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

product line

Vetec

assay

98%

mp

678-680 °C (lit.)

density

4 g/mL at 25 °C (lit.)

SMILES string

Cl[Pd]Cl

InChI

1S/2ClH.Pd/h2*1H;/q;;+2/p-2

InChI key

PIBWKRNGBLPSSY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Qiu Lin et al.
Environment international, 135, 105353-105353 (2019-12-13)
Tetrabromobisphenol A (TBBPA) has aroused serious pollution in surface sediment. To date, whether and how iron-based nanoparticles could stimulate TBBPA in situ anaerobic biodegradation in sediment remains poorly understood. In this study, the distinctly enhanced TBBPA degradation activity with the
A M Lacerda et al.
Nanoscale, 7(29), 12331-12335 (2015-07-08)
A photocatalyst consisting of nanostructured Pd photochemically deposited on 20 nm TiO2 displays a reaction half-life for rhodamine b decolourisation of 0.5 minute compared to the 9.4 minutes for unmodified P25 under identical reaction conditions. We associate this increased decolourisation
E Sutter et al.
Nature communications, 5, 4946-4946 (2014-09-12)
Galvanic replacement reactions provide an elegant way of transforming solid nanoparticles into complex hollow morphologies. Conventionally, galvanic replacement is studied by stopping the reaction at different stages and characterizing the products ex situ. In situ observations by liquid-cell electron microscopy
Guilue Guo et al.
Chemistry, an Asian journal, 9(9), 2555-2559 (2014-06-26)
The coupling of proteins with self-assembly properties and proteins that are capable of recognizing and mineralizing specific inorganic species is a promising strategy for the synthesis of nanoscale materials with controllable morphology and functionality. Herein, GPG-AG3 protein fibers with both
Sanki Asakawa et al.
PloS one, 10(3), e0119428-e0119428 (2015-03-06)
Nickel (Ni) ions easily elute from many alloys and elicit inflammation and allergies. Previous studies have shown that infections due to the implantation of medical devices cause inflammation and enhance the elution of Ni ions (Ni²⁺). However, cross-talk between infection-

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service