Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900825

Sigma-Aldrich

Tetrakis(triphenylphosphine)palladium(0)

Vetec, reagent grade, 98%

Synonym(s):

Palladium-tetrakis(triphenylphosphine), Pd(PPh3)4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd[(C6H5)3P]4
CAS Number:
Molecular Weight:
1155.56
Beilstein/REAXYS Number:
6704828
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

storage temp.

2-8°C

SMILES string

[Pd].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Pd/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h4*1-15H;

InChI key

NFHFRUOZVGFOOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhan Zhang et al.
Angewandte Chemie (International ed. in English), 53(34), 8980-8984 (2014-07-01)
tert-Butoxyacetylene is shown to undergo Sonogashira coupling with aryl iodides to yield aryl-substituted tert-butyl ynol ethers. These intermediates participate in a [1,5]-hydride shift, which results in the extrusion of isobutylene and the generation of aryl ketenes. The ketenes are trapped
Chanchal Chakraborty et al.
ACS applied materials & interfaces, 7(34), 19034-19042 (2015-08-19)
A platinum(II)-based, luminescent, metallo-supramolecular polymer (PolyPtL1) having an inherent dipole moment was synthesized via complexation of Pt(II) ions with an asymmetric ligand L1, containing terpyridyl and pyridyl moieties. The synthesized ligand and polymer were well characterized by various NMR techniques
Debabrata Jana et al.
Organic & biomolecular chemistry, 13(43), 10663-10674 (2015-09-09)
Four pyrene-vinyl-tetraphenylethylene based conjugated materials were synthesized and characterized by FT-IR, NMR, and mass spectroscopy. The photophysical (including absorption, fluorescence, and fluorescence lifetime) and aggregation properties in tetrahydrofuran were investigated. The photophysical and aggregation behavior depends on the spacer, substituent
Shouchun Yin et al.
Soft matter, 11(22), 4424-4429 (2015-05-01)
In this paper, a pyrene moiety is incorporated into a bolaamphiphile to form a novel molecule denoted PRB. Above the critical micelle concentration, PRB forms nanodisks in the aqueous solution. The addition of acetate ions induces a morphological change in
Haiyin Li et al.
Macromolecular bioscience, 14(10), 1361-1367 (2014-06-14)
Phospholipid monomer and aggregation-induced emission (AIE) dye-based dimers are incorporated via reversible addition-fragmentation transfer polymerization to afford cross-linked zwitterionic fluorescent copolymers. Such copolymers are prone to self-assembly into fluorescent polymeric nanoparticles (FPNs) in physiological solution due to their amphiphilic nature.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service