Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900824

Sigma-Aldrich

Palladium(II) acetylacetonate

Vetec, reagent grade, 98%

Synonym(s):

Palladium(II) 2,4-pentanedionate, Pd(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(C5H7O2)2
CAS Number:
Molecular Weight:
304.64
Beilstein/REAXYS Number:
4136188
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder

reaction suitability

core: aluminum

mp

200-251 °C (dec.)

SMILES string

CC(=O)\C=C(\C)O[Pd]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Pd/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

JKDRQYIYVJVOPF-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan W Zeng et al.
Physical chemistry chemical physics : PCCP, 20(37), 24389-24398 (2018-09-18)
Palladium nanoparticles were synthesized directly in solid fluoropolymer films by thermal decomposition of a palladium acetylacetonate precursor molecularly infused in the fluoropolymer matrix. This chemical infusion synthesis technique was studied using transmission electron microscopy along with selective area electron diffraction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service