Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900809

Sigma-Aldrich

N,N-Dimethylformamide

Vetec, reagent grade, anhydrous, ≥99.8%

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein/REAXYS Number:
605365
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
Pricing and availability is not currently available.

grade

anhydrous
reagent grade

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

product line

Vetec

assay

≥99.8%

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

refractive index

n20/D 1.430 (lit.)

pH

6.7

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solvent for many hydrophobic organic compounds.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

135.5 °F - closed cup

flash_point_c

57.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takaki Yamauchi et al.
Plant physiology, 169(1), 180-193 (2015-06-04)
In rice (Oryza sativa) roots, lysigenous aerenchyma, which is created by programmed cell death and lysis of cortical cells, is constitutively formed under aerobic conditions, and its formation is further induced under oxygen-deficient conditions. Ethylene is involved in the induction
Jixian Xu et al.
Nature communications, 6, 7081-7081 (2015-05-09)
Solution-processed planar perovskite devices are highly desirable in a wide variety of optoelectronic applications; however, they are prone to hysteresis and current instabilities. Here we report the first perovskite-PCBM hybrid solid with significantly reduced hysteresis and recombination loss achieved in
Samuel D Banister et al.
ACS chemical neuroscience, 6(8), 1445-1458 (2015-04-30)
Synthetic cannabinoid (SC) designer drugs featuring bioisosteric fluorine substitution are identified by forensic chemists and toxicologists with increasing frequency. Although terminal fluorination of N-pentyl indole SCs is sometimes known to improve cannabinoid type 1 (CB1) receptor binding affinity, little is
L Piazza et al.
Nature communications, 6, 6407-6407 (2015-03-03)
Surface plasmon polaritons can confine electromagnetic fields in subwavelength spaces and are of interest for photonics, optical data storage devices and biosensing applications. In analogy to photons, they exhibit wave-particle duality, whose different aspects have recently been observed in separate
S Ollier et al.
Journal of dairy science, 97(8), 4942-4954 (2014-06-03)
A cow's risk of acquiring a new intramammary infection during the dry period increases with milk production at drying off and decreases as mammary gland involution progresses. A method commonly used to reduce milk production is a drastic reduction in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service