Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900802

Sigma-Aldrich

Cesium carbonate

Vetec, reagent grade, 99%

Synonym(s):

Carbonic acid dicesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cs2CO3
CAS Number:
Molecular Weight:
325.82
Beilstein/REAXYS Number:
4546405
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

mp

610 °C (dec.) (lit.)

SMILES string

[Cs+].[Cs+].[O-]C([O-])=O

InChI

1S/CH2O3.2Cs/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

FJDQFPXHSGXQBY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Promotes the efficient O-alkylation of alcohols to form mixed alkyl carbonates.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Repr. 2 - STOT RE 2 Oral

target_organs

Kidney,Adrenal gland,Testes

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard C Burrell et al.
Journal of labelled compounds & radiopharmaceuticals, 57(10), 600-605 (2014-09-10)
Bristol-Myers Squibb and others are developing drugs that target novel mechanisms to combat Alzheimer's disease. γ-Secretase inhibitors are one class of potential therapies that have received considerable attention. (R)-2-(4-Chloro-N-(2-fluoro-4-(1,2,4-oxadiazol-3-yl)benzyl)phenylsulfonamido)-5,5,5-trifluoropentanamide (Avagacestat) is a γ-secretase-inhibiting drug that has been investigated by Bristol-Myers
Xi Zhang et al.
Small (Weinheim an der Bergstrasse, Germany), 11(45), 6091-6096 (2015-10-20)
A one-step method to produce ≈12 nm hydrodynamic diameter water-soluble CdSe/ZnS quantum dots (QDs), as well as CdS/ZnS, ZnSe/ZnMnS/ZnS, AgInS2 /ZnS, and CuInS2 /ZnS QDs, by ligand exchange with a near-monolayer of organosilane caps is reported. The method cross-links the
Ved Vati Singh et al.
Dalton transactions (Cambridge, England : 2003), 43(33), 12555-12563 (2014-07-10)
Nanoflowers and nanospheres of Pd4S have been prepared for the first time from a single source precursor complex, [PdCl2(PhS-CH2CH2CH2-NH2)] (), by its one pot thermolysis at 195 °C. In oleylamine, flower shaped nanoparticles of Pd4S were formed but in an
Ved Vati Singh et al.
Dalton transactions (Cambridge, England : 2003), 44(2), 725-732 (2014-11-20)
The metastable tetragonal Cu2Se phase as nanoflakes has been synthesized for the first time by treating CuCl2 taken in a mixture (1:1) of 1-octadecene and oleylamine with H2N-(CH2)3-SePh dissolved in 1-octadecene. Powder X-ray diffraction (PXRD), HRTEM, SEM-EDX and XPS have
Young-Shin Park et al.
ACS nano, 9(10), 10386-10393 (2015-08-28)
Lead-halide-based perovskites have been the subject of numerous recent studies largely motivated by their exceptional performance in solar cells. Electronic and optical properties of these materials have been commonly controlled by varying the composition (e.g., the halide component) and/or crystal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service